missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygram CEL 300 PEI/UV254 Polyester Sheets

Product Code. 10013021 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10013021 20 x 20 cm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10013021 Supplier Macherey-Nagel™ Supplier No. 801063
  • ONLINE EXCLUSIVE PRICE
    £215.31
    £191.34 / Pack of 25
    Save £23.97
    11% Off
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TLC polyester sheets with fibrous cellulose impregnated with polyethyleneimine (PEI).

Specifications

Type Polyester Sheet
Fluorescent Indicator UV254
Thickness (Metric) 0.1 mm
Dimensions (L x W) 20 x 20 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.