missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Polyethylene Industrial Grade Tape

Product Code. 13108585 Shop All Fisherbrand Products
Click to view available options
Color:
Orange
Red
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13108585

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Red
Material Polyethylene
Product Type Industrial Grade Tape
For Use With (Application) Aisle marking, labeling, batching, clean-build projects
Size (Metric) 33 m x 19 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.