missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol monomethyl ether p-toluenesulfonate, M.W. 5,000, Thermo Scientific Chemicals
£38.60 - £161.00
Chemical Identifiers
CAS | 58320-73-3 |
---|---|
Molecular Formula | C10H14O4S |
Molecular Weight (g/mol) | 230.278 |
InChI Key | TZXJJSAQSRHKCZ-UHFFFAOYSA-N |
Synonym | 2-methoxyethyl p-toluenesulfonate, p-toluenesulfonic acid 2-methoxyethyl ester, 2-methoxyethyl tosylate, ethanol, 2-methoxy-, 1-4-methylbenzenesulfonate, 2-methoxyethyltosylate, acmc-209e3c, aronis23919, 2-methoxyethyl-p-toluenesulfonate, 2-methoxyethyl p-toluenesulphonate, ethylene glycol methyl ether tosylate |
PubChem CID | 86977 |
IUPAC Name | 2-methoxyethyl 4-methylbenzenesulfonate |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)OCCOC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15487315
|
Thermo Scientific Alfa Aesar
046824.03 |
1 g |
£38.60
1g |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15497315
|
Thermo Scientific Alfa Aesar
046824.06 |
5 g |
£161.00
5g |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
58320-73-3 | |
230.278 | |
2-methoxyethyl p-toluenesulfonate, p-toluenesulfonic acid 2-methoxyethyl ester, 2-methoxyethyl tosylate, ethanol, 2-methoxy-, 1-4-methylbenzenesulfonate, 2-methoxyethyltosylate, acmc-209e3c, aronis23919, 2-methoxyethyl-p-toluenesulfonate, 2-methoxyethyl p-toluenesulphonate, ethylene glycol methyl ether tosylate | |
2-methoxyethyl 4-methylbenzenesulfonate |
C10H14O4S | |
TZXJJSAQSRHKCZ-UHFFFAOYSA-N | |
86977 | |
CC1=CC=C(C=C1)S(=O)(=O)OCCOC |
Specifications
White | |
Powder | |
2-methoxyethyl p-toluenesulfonate, p-toluenesulfonic acid 2-methoxyethyl ester, 2-methoxyethyl tosylate, ethanol, 2-methoxy-, 1-4-methylbenzenesulfonate, 2-methoxyethyltosylate, acmc-209e3c, aronis23919, 2-methoxyethyl-p-toluenesulfonate, 2-methoxyethyl p-toluenesulphonate, ethylene glycol methyl ether tosylate | |
CC1=CC=C(C=C1)S(=O)(=O)OCCOC | |
230.278 | |
Polyethylene glycol monomethyl ether p-toluenesulfonate, M.W. 5,000 |
58320-73-3 | |
C10H14O4S | |
TZXJJSAQSRHKCZ-UHFFFAOYSA-N | |
2-methoxyethyl 4-methylbenzenesulfonate | |
86977 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only