missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol monomethyl ether p-toluenesulfonate, M.W. 5,000

Product Code. 15497315
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15497315

Brand: Thermo Scientific Alfa Aesar 046824.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 58320-73-3 | C10H14O4S | 230.278 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58320-73-3
Molecular Formula C10H14O4S
Molecular Weight (g/mol) 230.278
InChI Key TZXJJSAQSRHKCZ-UHFFFAOYSA-N
Synonym 2-methoxyethyl p-toluenesulfonate, p-toluenesulfonic acid 2-methoxyethyl ester, 2-methoxyethyl tosylate, ethanol, 2-methoxy-, 1-4-methylbenzenesulfonate, 2-methoxyethyltosylate, acmc-209e3c, aronis23919, 2-methoxyethyl-p-toluenesulfonate, 2-methoxyethyl p-toluenesulphonate, ethylene glycol methyl ether tosylate
PubChem CID 86977
IUPAC Name 2-methoxyethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCOC

Specifications

Color White
Physical Form Powder
Quantity 5 g
Chemical Name or Material Polyethylene glycol monomethyl ether p-toluenesulfonate, M.W. 5,000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.