missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polyethylene Cap

Product Code. 15178592 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Transparent
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15178592 Transparent Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15178592 Supplier Macherey-Nagel™ Supplier No. 702129
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Closure Color Transparent
Color Transparent
Diameter (Metric) Outer 4.3 mm
Material Polyethylene
Septum Yes
For Use With (Application) Used as Washer Bottle Closures or Interim Closures
Includes With Septum
No. per Pack 100
Packaging Type 100 Pack
Closure Material Polyethylene
Quantity 100 Pk.
Thickness (Metric) 1.3 mm
Height (Metric) 9.1 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.