missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polycystin-1/PKD1 Antibody (PKS161f/B7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £337.00
Specifications
| Antigen | Polycystin-1/PKD1 |
|---|---|
| Clone | PKS161f/B7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642609
|
Novus Biologicals
NBP2-50247-0.025ml |
0.025 mL |
£169.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657498
|
Novus Biologicals
NBP2-50247 |
0.1 mL |
£337.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Polycystin-1/PKD1 Monoclonal antibody specifically detects Polycystin-1/PKD1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Polycystin-1/PKD1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Phospho Specific | |
| PBS | |
| 5310 | |
| IgG1 κ | |
| Protein A purified |
| PKS161f/B7 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Autosomal dominant polycystic kidney disease 1 protein, PBP, Pc-1, polycystic kidney disease 1 (autosomal dominant), polycystic kidney disease-associated protein, polycystin 1, polycystin-1, transient receptor potential cation channel, subfamily P, member 1, TRPP1 | |
| Recombinant protein (233 aa) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title