missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polycystin-1/PKD1 Antibody (PKS161f/B7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50247-0.025ml
This item is not returnable.
View return policy
Description
Polycystin-1/PKD1 Monoclonal antibody specifically detects Polycystin-1/PKD1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Polycystin-1/PKD1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Autosomal dominant polycystic kidney disease 1 protein, PBP, Pc-1, polycystic kidney disease 1 (autosomal dominant), polycystic kidney disease-associated protein, polycystin 1, polycystin-1, transient receptor potential cation channel, subfamily P, member 1, TRPP1 | |
| Recombinant protein (233 aa) | |
| 0.025 mL | |
| Phospho Specific | |
| 5310 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry | |
| PKS161f/B7 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction