Learn More
Kartell™ Polycarbonate/Polypropylene Pipette Tip
Make pipetting safer and simple for even the most sensitive applications. Kartell™ PC/PP Pipette Tips are compatable with a wide variety of pipette models.
Brand: Kartell™ 0960500
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Made of Polycarbonate/Polypropylene
- Tip for micropipette
- Supplied in rack
- Compatability varies
Specifications
Pipette tips | |
Yellow | |
Rack | |
Kartell™, Eppendorf™, Socorex™, Finnpipette™, Brand™, Hamilton™ | |
Polycarbonate/Polypropylene |
2 to 200 μL | |
CE/IVD | |
Universal | |
Pipettor Tip |