Learn More
R&D Systems™ Poly(ADP-ribose) Polymer/pADPr
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
- The PAR chain length ranges from 2 to 300 monomers
Specifications
Specifications
| Quantity | 100 μL |
| Product Type | Poly(ADP-ribose) (PAR) Polymer/pADPr |
| Content And Storage | Store the unopened product at -80° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |