missing translation for 'onlineSavingsMsg'
Learn More

POLR2D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£328.00

Specifications

Antigen POLR2D
Dilution Western Blot 0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18481481
View Documents
Novus Biologicals
NBP1-92280-25ul
25 μL
£328.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

POLR2D Polyclonal specifically detects POLR2D in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

POLR2D
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
DNA-directed RNA polymerase II 16 kDa polypeptide, DNA-directed RNA polymerase II subunit D, DNA-directed RNA polymerase II subunit RPB4, HSRBP4, HSRPB4, polymerase (RNA) II (DNA directed) polypeptide D, RBP4, RNA polymerase II 16 kDa subunit, RNA polymerase II subunit B4, RNA polymerase II subunit hsRBP4, RPB16
POLR2D
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Rabbit
Stem Cell Markers
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
5433
This antibody was developed against Recombinant Protein corresponding to amino acids:NRETIASVRSLLLQKKLHKFELACLANLCPETAEESKALIPSLEGRFEDEELQQILDDIQTKRSFQY
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.