missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ PolarSafe™ Cryogenic Storage Dewars with Racks

Cole-Parmer™ PolarSafe™ Cryogenic Storage Dewars with Racks safely store samples for up to 200 days without liquid nitrogen refill

Brand:  Cole-Parmer™ 4426058

Additional Details : Weight : 29.48350kg

 View more versions of this product

Product Code. 16649551

  • £2015.00 / Each
Estimated Shipment: 26-07-2024
to see stock.
Add to basket

Description

Description

  • Liquid nitrogen dewars available in sizes from 2 L to 115 L
  • Ultra low evaporation loss enhances cryogen protection
  • Durable aluminum construction
  • Dual handles aid mobility
  • 5 year vacuum warranty
Specifications

Specifications

12.7 cm
5 in.
Static Holding Time: 130 Days
Aluminum
55.88 cm
55.88 cm
22 in.
22 in.
47 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers