missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ PolarSafe™ Cryogenic Storage Dewars with Racks
Cole-Parmer™ PolarSafe™ Cryogenic Storage Dewars with Racks safely store samples for up to 200 days without liquid nitrogen refill
£485.00 - £6350.41
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
16659551
|
Cole-Parmer™
4426059 |
2 L |
£485.00
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
16629551
|
Cole-Parmer™
4426056 |
30 L |
£1423.49
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16639551
|
Cole-Parmer™
4426057 |
35 L |
£1594.22
Each |
In Stock
Log in to see stock. |
|||||
16649551
|
Cole-Parmer™
4426058 |
47 L |
£2015.00
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
16609551
|
Cole-Parmer™
4426054 |
95 L |
£6350.41
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
Description
- Liquid nitrogen dewars available in sizes from 2 L to 115 L
- Ultra low evaporation loss enhances cryogen protection
- Durable aluminum construction
- Dual handles aid mobility
- 5 year vacuum warranty
Specifications
Aluminum |