missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ PolarSafe™ Cryogenic Storage Dewars with Racks

Cole-Parmer™ PolarSafe™ Cryogenic Storage Dewars with Racks safely store samples for up to 200 days without liquid nitrogen refill

Brand:  Cole-Parmer™ 4426056

Additional Details : Weight : 15.42210kg

 View more versions of this product

Product Code. 16629551

  • £1423.49 / Each
Estimated Shipment: 16-09-2024
to see stock.
Add to basket

Description

Description

  • Liquid nitrogen dewars available in sizes from 2 L to 115 L
  • Ultra low evaporation loss enhances cryogen protection
  • Durable aluminum construction
  • Dual handles aid mobility
  • 5 year vacuum warranty
Specifications

Specifications

12.5 cm
4.875 in.
Static Holding Time: 71 Days
Aluminum
76.2 cm
76.2 cm
30 in.
30 in.
30 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers