missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (RTD4E10), mFluor Violet 450 SE, Novus Biologicals™
Hamster Monoclonal Antibody
£508.00
Specifications
| Antigen | Podoplanin |
|---|---|
| Clone | RTD4E10 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 450 SE |
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse, Human (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunoprecipitationSpecifications
| Podoplanin | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| mFluor Violet 450 SE | |
| Hamster | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| Murine thymic stromal cell line | |
| Primary |
| RTD4E10 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Human (Negative) | |
| 10630 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title