missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (RTD4E10), mFluor Violet 450 SE, Novus Biologicals™
Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP2-36058MFV450
This item is not returnable.
View return policy
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse, Human (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Spezifikation
| Podoplanin | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| Murine thymic stromal cell line | |
| 0.1 mL | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 10630 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| RTD4E10 | |
| 50 mM Sodium Borate | |
| Hamster | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, Human (Negative) | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur