Learn More
Thermo Scientific™ PocketTips™ Pipette Tips
Product Code. 12957217
Click to view available options
Volume (Metric):
250 nL
Unit Size:
Pack of 4800
Product Code. 12957217
Brand: Thermo Scientific™ FX96P20250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Eliminate time-consuming steps, reduce lab ware usage and accelerate assays with PocketTips™ Pipette Tips.
Eliminate time-consuming pipetting steps, reduce lab ware usage and accelerate assays such as PCR primer dilutions with user-friendly PocketTips™ Pipette Tips, which are compatible on multiple workstations. The external dimensions of the tip and rack are the same as standard automation tips, requiring minimal modification and set-up. Simple, economical assay miniaturization preserves compounds and reduces reagent costs.
Features:
  • Transfer samples directly to the assay plate
  • Combine sample and assay material with soft in-tip mixing
  • Coefficient of variation (CV) of less than 6% can be achieved for any pocket size
  • Color: Clear
Recommended for:
Cell-based or low DMSO tolerant assays, selectivity assays (P450), potency determinations (IC50), compound storage and delivery from compound libraries, protein and enzyme aqueous based assays, automated clinical diagnostics and kinetic read (FLIPR) assays.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 250 nL
Color Clear
Format Rack