missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PocketTip™ D.A.R.T.'S™ (Disposable Automation Research Tips)

Product Code. 11972271
Click to view available options
Volume (Metric):
100 nL
250 nL
50 nL
Sterility:
Non-sterile
Sterile
Packaging:
20 x 384 tips
20 x 96 tips
Unit Size:
Pack of 3840
Pack of 960
This item is not returnable. View return policy

Product Code. 11972271

missing translation for 'mfr': Thermo Scientific™ 5697250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing.

Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing. Thermo Scientific™ PocketTip™ D.A.R.T.'s™ achieve lower volume transfer capability due to a combination of the internally molded capillary pocket and the unique surface sealing of the tip.

Features:

  • Allows existing Thermo Scientific™ Matrix™ automated liquid handlers to accommodate low-volume dispensing for compound screening procedures
  • Requires only minor procedural adjustments to Thermo Scientific™ PlateMate™ and Thermo Scientific™ Hydra™ DT instrumentation
  • Eliminate aqueous dilution steps that can cause precipitation by transferring nanoliter volumes directly to assay plate.
  • Control consistent accuracy and precision during the molding process
Validation and Compatibility:
  • Purity and straightness validation are performed on compatible instrumentation to ensure only the highest quality.
  • With instrument compatibility range starting from the Matrix Hydra DT, on-the-fly dose response curves or assay development can be set up with simple Hydra DT programming.
  • Higher throughput needs can be met with the Matrix PlateMate 2 x 3 and Catalyst Express or Matrix PlateMate Plus instruments.
  • Sales specialists are available to assist with instrument programming and modifications or instrument validation of any Matrix automated liquid handler.
Ordering Information:
  • Available in 96- and 384- channel format with 50, 100 and 250nL pocket sizes
  • Sterile format available for sensitive liquid handling procedures
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 250 nL
Filtered Non-filtered
Sterility Sterile
Format Rack
Tip Style Automation
Packaging 20 x 96 tips
Disposable Single-use
For Use With (Equipment) Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus
Product Line PocketTip

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.