missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PNU 282987

Description
Highly selective α7 nAChR agonist (Ki = 26 nM) displaying negligible blockade of α1β1γδ and α3β4 nAChRs (IC50 ≥ 60 μM). Found to be inactive against a panel of 32 receptors at 1 μM, except 5-HT3 receptors (Ki = 930 nM).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 264.75 |
| Purity | >98% |
| Molecular Weight (g/mol) | 319.23 |
| Chemical Name or Material | PNU 282987 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?