accessibility menu, dialog, popup

UserName

NNHOClH2OHCl

CAS RN 123464-89-1

IUPAC Name: N-{1-azabicyclo[2.2.2]octan-3-yl}-4-chlorobenzamide hydrate hydrochloride
Synonyms: N-{1-azabicyclo[2.2.2]octan-3-yl}-4-chlorobenzamide hydrate hydrochloride
Molecular Weight (g/mol): 319.23
Molecular Formula: C14H20Cl2N2O2
InChi Key: OCWLMMBVXIESNP-UHFFFAOYNA-N
SMILES: O.Cl.ClC1=CC=C(C=C1)C(=O)NC1CN2CCC1CC2
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results