missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNR/NR2E3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00
Specifications
| Antigen | PNR/NR2E3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18743294
|
Novus Biologicals
NBP2-22466-0.025mg |
0.025 mg | |||||||
Description
PNR/NR2E3 Polyclonal specifically detects PNR/NR2E3 in Human, Rat samples. It is validated for Western Blot.Specifications
| PNR/NR2E3 | |
| Western Blot 2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Rat | |
| Nuclear receptor subfamily 2 group E member 3, nuclear receptor subfamily 2, group E, member 3, photoreceptor-specific nuclear receptor, PNRESCS, rd7, Retina-specific nuclear receptor, RNR, RP37MGC49976 | |
| NR2E3 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS and 1 mg/mL BSA with 0.05% Sodium Azide | |
| 10002 | |
| Synthetic peptide corresponding to residues R(392) K T I G N T P M E K L L C D M F K N(410) of human PNR. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title