missing translation for 'onlineSavingsMsg'
Learn More

PNR/NR2E3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-22466-0.025mg

 View more versions of this product

Product Code. 18743294

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

PNR/NR2E3 Polyclonal specifically detects PNR/NR2E3 in Human, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PNR/NR2E3
Polyclonal
Unconjugated
PBS and 1 mg/mL BSA with 0.05% Sodium Azide
NR2E3
Synthetic peptide corresponding to residues R(392) K T I G N T P M E K L L C D M F K N(410) of human PNR.
0.025 mg
Primary
Human, Rat
IgG
Western Blot
1 mg/mL
Western Blot 2 μg/mL
Nuclear receptor subfamily 2 group E member 3, nuclear receptor subfamily 2, group E, member 3, photoreceptor-specific nuclear receptor, PNRESCS, rd7, Retina-specific nuclear receptor, RNR, RP37MGC49976
Rabbit
Affinity Purified
RUO
10002
Store at -20°C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only