missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMT Antibody (CL11434), Novus Biologicals™
Mouse Monoclonal Antibody
£356.00 - £511.00
Specifications
| Antigen | PNMT |
|---|---|
| Clone | CL11434 |
| Dilution | Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620655
|
Novus Biologicals
NBP3-21193-100ul |
100 μg |
£511.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680844
|
Novus Biologicals
NBP3-21193-25ul |
25 μg |
£356.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PNMT Monoclonal antibody specifically detects PNMT in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| PNMT | |
| Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.1.1, EC 2.1.1.28, Noradrenaline N-methyltransferase, PENTMGC34570, phenylethanolamine N-methylase, phenylethanolamine N-methyltransferase, PNMTase | |
| This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of P11086. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL11434 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS, pH 7.2, 40% glycerol | |
| 5409 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title