missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMT Antibody (CL11434), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21193-25ul
This item is not returnable.
View return policy
Description
PNMT Monoclonal antibody specifically detects PNMT in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PNMT | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL11434 | |
| Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| EC 2.1.1, EC 2.1.1.28, Noradrenaline N-methyltransferase, PENTMGC34570, phenylethanolamine N-methylase, phenylethanolamine N-methyltransferase, PNMTase | |
| This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of P11086. Actual immunogen sequence is proprietary. | |
| 25 μg | |
| Cancer | |
| 5409 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction