missing translation for 'onlineSavingsMsg'
Learn More

PNKD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen PNKD
Concentration 0.31 mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16838763
View Documents
Proteintech
11584-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16828763
View Documents
Proteintech
11584-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Probable hydrolase that plays an aggravative role in the development of cardiac hypertrophy via activation of the NF-kappa-B signaling pathway.
Specifications

Specifications

PNKD
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Mouse, Human
Q69ZP3, Q8N490
100188944, 25953, 56695
PNKD Fusion Protein Ag2176
Primary
-20°C
PNKD
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
BRP17, DYT8, FKSG19, FPD1, KIAA1184, KIPP1184, MR 1, MR1, Myofibrillogenesis regulator 1, PDC, PNKD, Probable hydrolase PNKD, TAHCCP2, UNQ2491/PRO5778
PNKD
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.