missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNKD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11584-1-AP-20UL
This item is not returnable.
View return policy
Description
Probable hydrolase that plays an aggravative role in the development of cardiac hypertrophy via activation of the NF-kappa-B signaling pathway.Specifications
| PNKD | |
| Polyclonal | |
| Unconjugated | |
| PNKD | |
| BRP17, DYT8, FKSG19, FPD1, KIAA1184, KIPP1184, MR 1, MR1, Myofibrillogenesis regulator 1, PDC, PNKD, Probable hydrolase PNKD, TAHCCP2, UNQ2491/PRO5778 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100188944, 25953, 56695 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q69ZP3, Q8N490 | |
| PNKD | |
| PNKD Fusion Protein Ag2176 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction