missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNCK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-98426
This item is not returnable.
View return policy
Description
PNCK Polyclonal specifically detects PNCK in Rat samples. It is validated for Western Blot.
Specifications
| PNCK | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| BSTK3, calcium/calmodulin-dependent protein kinase type 1B, CaM kinase I beta, CaM kinase IB, CaMK1b, CaM-KI beta, CaMKI-beta, EC 2.7.11, EC 2.7.11.17, FLJ50403, FLJ50549, FLJ56451, FLJ59811, MGC45419, pregnancy up-regulated non-ubiquitously expressed CaM kinase, pregnancy upregulated non-ubiquitously expressed CaM kinase, Pregnancy up-regulated non-ubiquitously-expressed CaM kinase | |
| Rabbit | |
| 37 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_058971 | |
| PNCK | |
| The immunogen for this antibody is Pnck - C-terminal region. Peptide sequence QKNFARTHWKRAFNATSFLRHIRKLGQSPEGEEASRQGMTRHSHPGLGTS. | |
| Affinity purified | |
| RUO | |
| 139728 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction