missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMS2 Recombinant Rabbit Monoclonal Antibody (BLR142H)

Rabbit Recombinant Monoclonal Antibody
£128.00 - £321.00
Specifications
| Antigen | PMS2 |
|---|---|
| Clone | BLR142H |
| Concentration | 10 μg/mL |
| Content And Storage | 4°C |
| Applications | Immunoprecipitation, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17775839
![]() |
Invitrogen™
MA544471 |
100 μL |
£321.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17785839
|
Invitrogen™
MA544472 |
20 μL |
£128.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PMS2 Recombinant Monoclonal Antibody for Western Blot, IP
PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.Specifications
| PMS2 | |
| 10 μg/mL | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
| P54278 | |
| 5395 | |
| Peptide representing a region between residue 550 and 600 of Human mismatch repair endonuclease PMS2. | |
| Antibody |
| BLR142H | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| PMS2 | |
| AW555130; DNA mismatch repair protein PMS2; H_DJ0042M02.9; HNPCC4; mismatch repair endonuclease PMS2; MLH4; PMS1 homolog 2, mismatch repair protein; PMS1 homolog 2, mismatch repair system component; PMS1 protein homolog 2; PMS2; PMS2 postmeiotic segregation increased 2; PMS2 postmeiotic segregation increased 2 (S. cerevisiae); PMS2CL; PMSL2; postmeiotic segregation increased 2; postmeiotic segregation increased 2 (S. cerevisiae); postmeiotic segregation increased 2 nirs variant 6 | |
| PMS2 | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title