missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMS2 Recombinant Rabbit Monoclonal Antibody (BLR142H)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544471
This item is not returnable.
View return policy
Description
PMS2 Recombinant Monoclonal Antibody for Western Blot, IP
PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.
Specifications
| PMS2 | |
| Recombinant Monoclonal | |
| 10 μg/mL | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
| P54278 | |
| PMS2 | |
| Peptide representing a region between residue 550 and 600 of Human mismatch repair endonuclease PMS2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| BLR142H | |
| Unconjugated | |
| PMS2 | |
| AW555130; DNA mismatch repair protein PMS2; H_DJ0042M02.9; HNPCC4; mismatch repair endonuclease PMS2; MLH4; PMS1 homolog 2, mismatch repair protein; PMS1 homolog 2, mismatch repair system component; PMS1 protein homolog 2; PMS2; PMS2 postmeiotic segregation increased 2; PMS2 postmeiotic segregation increased 2 (S. cerevisiae); PMS2CL; PMSL2; postmeiotic segregation increased 2; postmeiotic segregation increased 2 (S. cerevisiae); postmeiotic segregation increased 2 nirs variant 6 | |
| Rabbit | |
| Purified | |
| RUO | |
| 5395 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction