Learn More
Bel-Art™ Scienceware™ PMP Urbanti High-speed Filter Paper
Unsurpassed for non-vacuum, high-speed filtration
Brand: Bel-Art™ Scienceware™ F14642-0000
Code : 02
Additional Details : Weight : 0.07708kg
Description
Unsurpassed for non-vacuum, high-speed filtration, these clear funnels have internal helicoid ribs to increase the filtration speed of filter paper. Vertical outer ribs prevent airlock. Unaffected by corrosive liquids and heat resistant to 177°C (350°F).Specifications
7.9cm | |
0.8cm | |
20.6 cm |
76 mL | |
PMP | |
Yes |