missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ PMP Griffin Beaker

Product Code. 10210891
Change view
Click to view available options
Capacity (Metric):
25 mL
100 mL
150 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 12
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10210891 150 mL Each
12783399 25 mL Pack of 12
12703409 100 mL Pack of 12
12723409 500 mL Pack of 6
12733409 1000 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10210891 Supplier VITLAB™ Supplier No. 60903

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Make your job easier and safer while handling a wide range of liquids with these polymethylpentene beakers. VITLAB™ PMP Griffin Beakers feature an easy to read, embossed red scale.

  • Crystal clear transparent graduated beakers
  • For autoclaving, models with raised graduations are recommended
  • To preserve markings, clean at no higher than 60°C

Specifications

Color Crystal Clear
Capacity (Metric) 150 mL
Material PMP
For Use With (Application) For Volume Measurement
Height (Metric) 80 mm
Type Griffin Beaker
Autoclavable Autoclavable
Graduated Yes
Certifications/Compliance ISO 7056
Scale Red Printed Scale
Style Griffin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.