missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PML Protein Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£176.00 - £318.00
Specifications
| Antigen | PML Protein |
|---|---|
| Concentration | 0.14 mg/ml |
| Dilution | Western Blot 1:750, ELISA 1:100-1:200, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18369906
|
Novus Biologicals
NBP3-18686-25UL |
25 μg |
£176.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18336134
|
Novus Biologicals
NBP3-18686-50UG |
50 μg |
£318.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PML Protein Polyclonal antibody specifically detects PML Protein in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| PML Protein | |
| Western Blot 1:750, ELISA 1:100-1:200, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| P29590 | |
| 5371 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| 0.14 mg/ml | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Zinc Finger | |
| PBS | |
| MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19 | |
| Polyclonal antibody raised in rabbit against human PML Protein (Promyelocytic leukemia), using 3 different KLH-conjugated synthetic peptides. | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title