missing translation for 'onlineSavingsMsg'
Learn More

PML Protein Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18369906 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μg
50 μg
Unit Size:
25µL
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369906 25 μg 25µL
18336134 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18369906 Supplier Novus Biologicals Supplier No. NBP31868625UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PML Protein Polyclonal antibody specifically detects PML Protein in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PML Protein
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.14 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:750, ELISA 1:100-1:200, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS
Gene Accession No. P29590
Gene Alias MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human PML Protein (Promyelocytic leukemia), using 3 different KLH-conjugated synthetic peptides.
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cancer, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 5371
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.