missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PML Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 33 publications
£169.00 - £499.00
Especificaciones
| Antigen | PML Protein |
|---|---|
| Concentration | 1.0 mg/mL |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|
18008963
|
Novus Biologicals
NB100-59787SS |
0.025 mg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18771424
|
Novus Biologicals
NB100-59787 |
£499.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
PML Protein Polyclonal specifically detects PML Protein in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Proximity Ligation Assay, Knockdown Validated.Specifications
| PML Protein | |
| Polyclonal | |
| Rabbit | |
| Cancer, Zinc Finger | |
| P29590 | |
| 5371 | |
| The immunogen recognized by this antibody maps to a region between residue 375 and 425 of promyelocytic leukemia using the numbering given in entry NP_150241.2 | |
| Primary | |
| Store at 4C. Do not freeze. |
| 1.0 mg/mL | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19 | |
| PML | |
| IgG | |
| Affinity Purified | |
| Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee, Primates, White-tufted-ear marmoset, Pygmy chimpanzee and Bornean orangutan. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title