missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PML Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 33 publications
Brand: Novus Biologicals NB100-59787SS
This item is not returnable.
View return policy
Description
PML Protein Polyclonal specifically detects PML Protein in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Proximity Ligation Assay, Knockdown Validated.
Specifications
| PML Protein | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee, Primates, White-tufted-ear marmoset, Pygmy chimpanzee and Bornean orangutan. | |
| Store at 4C. Do not freeze. |
| Immunoblot | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Flow Cytometry, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:200-1:1000, Immunoblotting, Proximity Ligation Assay 1:200-1:5000, KnockDown Validated | |
| P29590 | |
| PML | |
| The immunogen recognized by this antibody maps to a region between residue 375 and 425 of promyelocytic leukemia using the numbering given in entry NP_150241.2 | |
| 0.025 mg | |
| Cancer, Zinc Finger | |
| 5371 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction