missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLSCR1 Recombinant Rabbit Monoclonal Antibody (24GB1550)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen PLSCR1
Clone 24GB1550
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275657
GREENER_CHOICE
View Documents
Invitrogen™
MA557580
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30274905
View Documents
Invitrogen™
MA557581
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PLSCR1 Recombinant Monoclonal Antibody for Western Blot, Flow

The calcium-dependent plasma membrane protein phospholipid scramblase 1 (PLSCR1) contributes to the transbilayer movement of phosphatidylserine and other membrane phospholipids upon influx of calcium into the cytosol. This movement results in plasma membrane phospholipid remodelling and surface exposure of phosphatidylserine in injured or apoptotic cells, which leads to cell death. Interferons and other cytokines induce expression of PLSCR1, implying that PLSCR1 also functions in cytokine signaling pathways. EGF stilmulation results in tyrosine phosphorylation of PLSCR1 on tyrosines 69 and 74, which allows it to interact with Shc, and thereby connecting Src kinase activation to stimulation of the EGF receptor.
Specifications

Specifications

PLSCR1
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
PLSCR1
ca(2+)-dependent phospholipid scramblase 1; Erythrocyte phospholipid scramblase; MmTRA1a; MmTRA1b; NOR1; Phospholipid scramblase 1; phospholipid scramblase 2; PL scramblase 1; Plscr1; Tra1; Tra1a; Tra1b; transplantability-associated protein 1; Tras1; Tras2
PLSCR1
Primary
Affinity chromatography
24GB1550
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O15162, P58195, Q9JJ00
117540, 22038, 5359
A synthesized peptide derived from human Scramblase 1 (aa 20-55).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.