missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Plonmarlimab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen Plonmarlimab Humanized
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30282619
GREENER_CHOICE
View Documents
Invitrogen™
MA559681
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30284663
View Documents
Invitrogen™
MA559682
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Plonmarlimab, also known as TJM2 or TJ003234, is a humanized IgG1 monoclonal antibody that targets and neutralizes granulocyte-macrophage colony-stimulating factor (GM-CSF), a cytokine implicated in inflammatory and autoimmune conditions. CSF2, also known as GM-CSF (Granulocyte-Macrophage Colony-Stimulating Factor), is located on chromosome 5q31. This gene encodes a cytokine that functions as a growth factor for hematopoietic progenitor cells, influencing the proliferation, differentiation, and survival of white blood cells such as granulocytes and macrophages. The CSF2 gene plays a critical role in the immune response by stimulating the production of granulocytes and macrophages from progenitor cells in the bone marrow, thus aiding in the defense against infections and in inflammatory responses. CSF2 is involved in various physiological processes and pathological conditions, including its overexpression in certain cancers and autoimmune diseases. The CSF2 receptor is a heterodimer composed of an alpha subunit (specific for GM-CSF) and a beta subunit shared with other cytokine receptors. Additionally, GM-CSF has been found to modulate inflammatory responses in the central nervous system, particularly influencing microglial cells and playing a role in neuroinflammatory and neurodegenerative disorders.
Specifications

Specifications

Plonmarlimab Humanized
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.