Learn More
Invitrogen™ Plonmarlimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559682
 View more versions of this product
Product Code. 30284663
  • £1002.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Plonmarlimab, also known as TJM2 or TJ003234, is a humanized IgG1 monoclonal antibody that targets and neutralizes granulocyte-macrophage colony-stimulating factor (GM-CSF), a cytokine implicated in inflammatory and autoimmune conditions. CSF2, also known as GM-CSF (Granulocyte-Macrophage Colony-Stimulating Factor), is located on chromosome 5q31. This gene encodes a cytokine that functions as a growth factor for hematopoietic progenitor cells, influencing the proliferation, differentiation, and survival of white blood cells such as granulocytes and macrophages. The CSF2 gene plays a critical role in the immune response by stimulating the production of granulocytes and macrophages from progenitor cells in the bone marrow, thus aiding in the defense against infections and in inflammatory responses. CSF2 is involved in various physiological processes and pathological conditions, including its overexpression in certain cancers and autoimmune diseases. The CSF2 receptor is a heterodimer composed of an alpha subunit (specific for GM-CSF) and a beta subunit shared with other cytokine receptors. Additionally, GM-CSF has been found to modulate inflammatory responses in the central nervous system, particularly influencing microglial cells and playing a role in neuroinflammatory and neurodegenerative disorders.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plonmarlimab Humanized
Recombinant Monoclonal
Unconjugated
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A
RUO
Human
Antibody
IgG1