missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHO2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | PLEKHO2 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PLEKHO2 Polyclonal specifically detects PLEKHO2 in Mouse samples. It is validated for Western Blot.Specifications
| PLEKHO2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| DKFZp761K2312, FLJ38884, PH domain-containing family O member 2, PH domain-containing family Q member 1, PH domain-containing protein, pleckstrin homology domain containing, family O member 2, pleckstrin homology domain containing, family Q member 1, pleckstrin homology domain-containing family O member 2, Pleckstrin homology domain-containing family Q member 1, PLEKHQ1, PP1628, pp9099 | |
| The immunogen is a synthetic peptide directed towards the middle region of mouse PLEKHO2 (NP_694759.1). Peptide sequence LLRSPGNKVSDIKFQAPSGEEKESWIKALNEGINRGKNKAFDEVKVDKTC | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 80301 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title