missing translation for 'onlineSavingsMsg'
Learn More

PLC-beta 1 Antibody (S03-6E6), Novus Biologicals™

Rabbit Monoclonal Antibody

£328.00 - £500.00

Specifications

Antigen PLC-beta 1
Clone S03-6E6
Concentration 0.3 mg/mL
Dilution Western Blot 1:1000
Applications Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18627943
View Documents
Novus Biologicals
NBP3-19739-100ul
100 μg
£500.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18639043
View Documents
Novus Biologicals
NBP3-19739-50ul
50 μg
£328.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PLC-beta 1 Monoclonal antibody specifically detects PLC-beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

PLC-beta 1
0.3 mg/mL
Western Blot
Unconjugated
Rabbit
Lipid and Metabolism, Wnt Signaling Pathway
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
23236
IgG
Affinity purified
S03-6E6
Western Blot 1:1000
Monoclonal
Purified
RUO
Human, Mouse, Rat
EC 3.1.4.11, EIEE12, FLJ45792, inositoltrisphosphohydrolase, KIAA05811-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta 1, monophosphatidylinositol phosphodiesterase, phosphoinositidase C, Phosphoinositide phospholipase C-beta-1, phospholipase C, beta 1 (phosphoinositide-specific), Phospholipase C-beta-1, Phospholipase C-I, PI-PLC, PLC-154, PLC154,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-1, PLCB1A, PLCB1B, PLC-beta-1, PLC-I1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, triphosphoinositide phosphodiesterase
Recombinant protein of mouse PLC-beta 1 (Uniprot # Q9Z1B3)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.