missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLC-beta 1 Antibody (S03-6E6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19739-100ul
This item is not returnable.
View return policy
Description
PLC-beta 1 Monoclonal antibody specifically detects PLC-beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PLC-beta 1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot 1:1000 | |
| EC 3.1.4.11, EIEE12, FLJ45792, inositoltrisphosphohydrolase, KIAA05811-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta 1, monophosphatidylinositol phosphodiesterase, phosphoinositidase C, Phosphoinositide phospholipase C-beta-1, phospholipase C, beta 1 (phosphoinositide-specific), Phospholipase C-beta-1, Phospholipase C-I, PI-PLC, PLC-154, PLC154,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-1, PLCB1A, PLCB1B, PLC-beta-1, PLC-I1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, triphosphoinositide phosphodiesterase | |
| Recombinant protein of mouse PLC-beta 1 (Uniprot # Q9Z1B3) | |
| 100 μg | |
| Lipid and Metabolism, Wnt Signaling Pathway | |
| 23236 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S03-6E6 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction