Learn More
Platinum, 10% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals
£137.00 - £1620.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11304447
|
Thermo Scientific Alfa Aesar
038325.06 |
5 g |
£137.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314447
|
Thermo Scientific Alfa Aesar
038325.14 |
25 g |
£538.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324447
|
Thermo Scientific Alfa Aesar
038325.22 |
100 g |
£1620.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Platinum is used in catalytic converters, laboratory equipment, electrical application and electrodes, platinum resistance thermometers, dentistry equipment, and jeweler. It is also used in glass fiber applications, chemical processing, off-road vehicles and fuel cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Aromatic ring hydrogenation, aliphatic aldehydes and ketones to alcohols, conversion of -CH=CH- to -CH2-CH2-
Chemical Identifiers
7440-6-6 | |
MFCD00011179 |
Specifications
7440-6-6 | |
MFCD00011179 | |
Platinum, 10% on activated carbon powder, Standard, reduced, nominally 50% water wet |
Odorless | |
Soluble in aqua regia.. Insoluble in water and acid. |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only