missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 10% on activated carbon powder, standard, reduced, nominally 50% water wet

Product Code. 11304447
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304447 5 g 5g
11314447 25 g 25g
11324447 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11304447 Supplier Thermo Scientific Alfa Aesar Supplier No. 038325.06
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7440-6-6

Platinum is used in catalytic converters, laboratory equipment, electrical application and electrodes, platinum resistance thermometers, dentistry equipment, and jeweler. It is also used in glass fiber applications, chemical processing, off-road vehicles and fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Aromatic ring hydrogenation, aliphatic aldehydes and ketones to alcohols, conversion of -CH=CH- to -CH2-CH2-
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-6-7440
MDL Number MFCD00011179

Specifications

Odor Odorless
Quantity 5 g
Solubility Information Soluble in aqua regia.. Insoluble in water and acid.
Chemical Name or Material Platinum, 10% on activated carbon powder, Standard, reduced, nominally 50% water wet

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.