missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plate Count Agar

Product Code. 13255349
Change view
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
Plate Count Agar
Manufacturing Origin:
Germany
United Kingdom
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Media Type Manufacturing Origin Unit Size
13255349 10/Pk. 90mm Monoplate Plate Count Agar Germany Pack of 10
11973612 10/Pk. 90mm Monoplate Plate Count Agar United Kingdom Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13255349 Supplier Thermo Scientific™ Supplier No. PO5013A
  • ONLINE EXCLUSIVE PRICE
    £14.15
    £13.44 / Pack of 10
    Save £0.71
    5% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enumerate viable organisms in milk, water, food and dairy products using this non-selective medium.

Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Plate Count Agar.

This standard medium meets the formulation of APHA, AOAC and PHLS.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Tested in accordance with CEN ISO/TS 11133-2.
Color Ivory, transparent
Content And Storage 2° to 12°C
Format 90mm Monoplate
Manufacturing Origin Germany
Media Type Plate Count Agar
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.