missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plastin L Antibody (29G10), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Plastin L |
|---|---|
| Clone | 29G10 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30023995
|
Novus Biologicals
NBP3-26174-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028383
|
Novus Biologicals
NBP3-26174-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plastin L Monoclonal antibody specifically detects Plastin L in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Plastin L | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CP64, DKFZp781A23186, FLJ25423, FLJ26114, FLJ39956, LC64PPLS2bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)), LCP-1, LPL, L-plastin, L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P), Lymphocyte cytosolic protein 1, lymphocyte cytosolic protein 1 (L-plastin), Lymphocyte cytosolic protein-1 (plasmin), plastin 2, plastin-2 | |
| A synthesized peptide derived from Human Plastin L [UniProt P13796] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 29G10 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3936 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title