missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plastin L Antibody (29G10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26174-100ul
This item is not returnable.
View return policy
Description
Plastin L Monoclonal antibody specifically detects Plastin L in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| Plastin L | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 29G10 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| CP64, DKFZp781A23186, FLJ25423, FLJ26114, FLJ39956, LC64PPLS2bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)), LCP-1, LPL, L-plastin, L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P), Lymphocyte cytosolic protein 1, lymphocyte cytosolic protein 1 (L-plastin), Lymphocyte cytosolic protein-1 (plasmin), plastin 2, plastin-2 | |
| A synthesized peptide derived from Human Plastin L [UniProt P13796] | |
| 100 μL | |
| Cancer, Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction | |
| 3936 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction