Learn More
Saint-Gobain Plastic Sanitary Couplings
Cole-Parmer™ Sanitary Coupling with Barb, Polypropylene has a tapered funnel design to provide a steady fluid flow.
£71.30
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
12409650
|
Saint-Gobain
31512-01 |
Polypropylene |
£71.30
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
11749769
|
Saint-Gobain
31512-02 |
Polypropylene |
£71.30
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
Description
- Reduces dead space and improves flow.
- Tapered funnel design provides a steady fluid flow.
- Smooth inner surface prevents particle entrapment.
- Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth.
- Material meets USP Class VI requirements and is FDA compliant for food contact.
Specifications
Polypropylene |