Learn More
Saint-Gobain Plastic Sanitary Couplings
Cole-Parmer™ Sanitary Coupling with Barb, Polypropylene has a tapered funnel design to provide a steady fluid flow.
Brand: Saint-Gobain 31512-02
Code : XX
Additional Details : Weight : 0.80000kg
Description
- Reduces dead space and improves flow.
- Tapered funnel design provides a steady fluid flow.
- Smooth inner surface prevents particle entrapment.
- Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth.
- Material meets USP Class VI requirements and is FDA compliant for food contact.
Specifications
Meets USP Class VI requirements and is FDA compliant for food contact | |
Polypropylene | |
FDA compliant for food contact | |
Gamma | |
1 13/16 in. |
Autoclavable | |
1/2 in. ID tubing, 1/2 in. mini clamp | |
Fits 1/2 in. ID tubing | |
31/32 in. |