Learn More
Edwards Vacuum™ Plastic Exhaust Nozzle Kit
Click to view available options
Type:
Exhaust Nozzle Kit
Unit Size:
Each
Specifications
Specifications
| Description | 0.75 in. to 15 mm |
| For Use With | nXDS6i, nXDS10i and nXDS15i Dry Scroll Pumps |
| Type | Exhaust Nozzle Kit |