missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasmolipin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00
Specifications
| Antigen | Plasmolipin |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18783294
|
Novus Biologicals
NBP2-22465-0.025mg |
0.025 mg | |||||||
Description
Plasmolipin Polyclonal specifically detects Plasmolipin in Rat samples. It is validated for Western Blot.Specifications
| Plasmolipin | |
| Western Blot 2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Rat | |
| Plasma membrane proteolipid, plasmolipin, PMLPTM4SF11plasma membrane proteolipid (plasmolipin), transmembrane 4 superfamily member 11 (plasmolipin) | |
| PLLP | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS and 1 mg/mL BSA with 0.05% Sodium Azide | |
| 51090 | |
| Synthetic peptides corresponding to residues C D(127) L T S L R G (S/T) R P Y N Q R S(141) of human and rat Plasmolipin. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title