missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasmolipin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22465-0.025mg
This item is not returnable.
View return policy
Description
Plasmolipin Polyclonal specifically detects Plasmolipin in Rat samples. It is validated for Western Blot.
Specifications
| Plasmolipin | |
| Polyclonal | |
| Unconjugated | |
| PBS and 1 mg/mL BSA with 0.05% Sodium Azide | |
| PLLP | |
| Synthetic peptides corresponding to residues C D(127) L T S L R G (S/T) R P Y N Q R S(141) of human and rat Plasmolipin. | |
| 0.025 mg | |
| Primary | |
| Rat | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2 μg/mL | |
| Plasma membrane proteolipid, plasmolipin, PMLPTM4SF11plasma membrane proteolipid (plasmolipin), transmembrane 4 superfamily member 11 (plasmolipin) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 51090 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction